Merck
  • An assessment of udp-glucuronosyltransferase induction using primary human hepatocytes.
An assessment of udp-glucuronosyltransferase induction using primary human hepatocytes.
Drug metabolism and disposition: the biological fate of chemicals (2004-01-08)
Matthew G Soars, David M Petullo, James A Eckstein, Steve C Kasper, Steven A Wrighton
ABSTRACT
Uridine diphosphate glucuronosyltransferases (UGTs) catalyze the glucuronidation of a wide range of xenobiotics and endogenous substrates. However, there is a lack of information concerning the response of human UGTs to inducers, and this observation prompted the current investigation. The glucuronidation of estradiol (3- and 17-positions), naphthol, propofol, and morphine (3- and 6-positions) was assessed against a battery of recombinant human UGTs to determine selective glucuronidation reactions for induction studies. The potential induction of the glucuronidation of estradiol at the 3-position, naphthol, propofol, and morphine at the 3-position was subsequently investigated in cultured primary human hepatocytes against a range of prototypic inducers including dexamethasone, 3-methylcholanthrene (3-MC), phenobarbital, rifampicin, and omeprazole. Treatment with 3-MC induced estradiol-3-glucuronidation (up to 2.5-fold) in four of five donors investigated. Statistically significant increases in naphthol glucuronidation (up to 1.7-fold) were observed following treatment with carbamazepine. UGT1A9-mediated propofol glucuronidation was induced by phenobarbital (up to 2.2-fold) and rifampicin (up to 1.7-fold). However, treatment with alpha-naphthoflavone and tangeretin resulted in a decrease in propofol glucuronidation (30% of control values). Statistically significant induction of morphine-3-glucuronidation was observed in at least three donors following treatment with phenobarbital, rifampicin, and carbamazepine. Each UGT isoform investigated displayed a distinct induction profile. Although statistically significant increases in glucuronidation were observed for each reaction studied, the level of induction was less than that observed for CYP1A2 or CYP3A4 and exhibited a large interdonor variability. The clinical relevance of the induction responses obtained in this study is unclear.
Product Description
Supelco
6β-Hydroxytestosterone solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
6β-Hydroxytestosterone, ≥97% (HPLC)