Merck
  • Primary causes of decreased mitochondrial oxygen consumption during metabolic depression in snail cells.
Primary causes of decreased mitochondrial oxygen consumption during metabolic depression in snail cells.
American journal of physiology. Regulatory, integrative and comparative physiology (2002-01-17)
Tammie Bishop, Julie St-Pierre, Martin D Brand
ABSTRACT
Cells isolated from the hepatopancreas of estivating snails (Helix aspersa) have strongly depressed mitochondrial respiration compared with controls. Mitochondrial respiration was divided into substrate oxidation (which produces the mitochondrial membrane potential) and ATP turnover and proton leak (which consume it). The activity of substrate oxidation (and probably ATP turnover) decreased, whereas the activity of proton leak remained constant in estivation. These primary changes resulted in a lower mitochondrial membrane potential in hepatopancreas cells from estivating compared with active snails, leading to secondary decreases in respiration to drive ATP turnover and proton leak. The respiration to drive ATP turnover and proton leak decreased in proportion to the overall decrease in mitochondrial respiration, so that the amount of ATP turned over per O2 consumed remained relatively constant and aerobic efficiency was maintained in this hypometabolic state. At least 75% of the total response of mitochondrial respiration to estivation was caused by primary changes in the kinetics of substrate oxidation, with only 25% or less of the response occurring through primary effects on ATP turnover.
Sigma-Aldrich
Methyltriphenylphosphonium bromide, 98%
Pricing and availability is not currently available.