Merck
Distribution of organochlorine pesticides in soils from South Korea.
Chemosphere (2001-04-12)
J H Kim, A Smith
ABSTRACT
Soil samples were collected from rice growing and industrial areas in South Korea and analysed for organochlorine pesticide content using gas chromatography with electron capture detection. The soils were monitored for the presence of 18 organochlorine pesticides. The main pesticides found were gamma- and delta-hexachlorocyclohexane, heptachlor epoxide and dieldrin. The range of concentrations, for each compound, was respectively, 0.17-0.94, 0.77-2.97, 1.38-48.0 and 0.32-0.49 ng/g soil. The highest values were found in soil obtained from rice fields indicating that, although the use of organochlorine pesticides has been discontinued since 1980, substantial concentrations of residues particularly the oxidised form of heptachlor remain in the soil.
Product Description
Supelco
δ-BHC, certified reference material, 1000 μg/mL in methanol
Supelco
δ-HCH, PESTANAL®, analytical standard