Merck
  • Mitochondrial phospholipid hydroperoxide glutathione peroxidase inhibits the release of cytochrome c from mitochondria by suppressing the peroxidation of cardiolipin in hypoglycaemia-induced apoptosis.
Mitochondrial phospholipid hydroperoxide glutathione peroxidase inhibits the release of cytochrome c from mitochondria by suppressing the peroxidation of cardiolipin in hypoglycaemia-induced apoptosis.
The Biochemical journal (2000-09-22)
K Nomura, H Imai, T Koumura, T Kobayashi, Y Nakagawa
ABSTRACT
Cytochrome c (cyt. c) is a proapoptotic factor that binds preferentially to cardiolipin (CL), a mitochondrial lipid, but not to cardiolipin hydroperoxide (CL-OOH). Cyt. c that had bound to CL liposomes was liberated on peroxidation of the liposomes by a radical. The generation of CL-OOH in mitochondria occurred before the release of cyt. c in rat basophile leukaemia (RBL)2H3 cells that had been induced to undergo apoptosis by exposure to hypoglycaemia with 2-deoxyglucose (2DG). The amount of cyt. c bound to CL prepared from the mitochondria of 2DG-treated cells was lower than that of untreated cells. The release of cyt. c was completely suppressed when the production of CL-OOH in mitochondria was inhibited by the overexpression of mitochondrial phospholipid hydroperoxide glutathione peroxidase (PHGPx). The fluorescence from CL-labelling dye (10-N-nonyl Acridine Orange) decreased on the induction of apoptosis by 2DG. However, no decrease in fluorescence was observed in PHGPx-overexpressing cells. Cyt. c was released from mitochondria that had been isolated from control cells on peroxidation by t-butylhydroperoxide, but no similar liberation of cyt. c from mitochondria isolated from mitochondrial PHGPx-overexpressing cells was observed. These findings suggest that the generation of CL-OOH in mitochondria might be a primary event that triggers the release of cyt. c from mitochondria in the apoptotic process in which mitochondrial PHGPx participates as an anti-apoptotic factor by preventing the formation of CL-OOH.
Sigma-Aldrich
Diethyl malate, ≥97%, FG