Merck
  • Minimization of cobalt nuclide emissions in supercritical water oxidation of spent resin.
Minimization of cobalt nuclide emissions in supercritical water oxidation of spent resin.
Chemosphere (2000-02-09)
Y J Huang, H P Wang, C T Li, Y C Chien
ABSTRACT
A rapid and complete destruction of organics in the supercritical water oxidation (SCWO) of the Co-exchanged resin was found experimentally. Due to an extremely low solubility of CoSO4 salt formed and separated effectively in the SCWO process, a minimal release of the nuclide Co would be warranted. In addition, recycling of Co nuclides is also possible by decomposition of the CoSO4 species at elevated temperatures (> 1040 K).
Sigma-Aldrich
Cobalt(II) sulfate heptahydrate, ReagentPlus®, ≥99%