Merck
  • Selective inhibition of inducible nitric oxide synthase maintains haemodynamic stability without untoward consequences for hepatic function or morphology.
Selective inhibition of inducible nitric oxide synthase maintains haemodynamic stability without untoward consequences for hepatic function or morphology.
The European journal of surgery = Acta chirurgica (2000-01-15)
Y Gundersen, T Saetre, T Scholz, T Hovig, P Lilleaasen, A O Aasen
ABSTRACT
To examine the effects of the inducible nitric oxide synthase inhibitor aminoethyl-isothiourea (AE-ITU) on haemodynamic measurements, and correlate these with hepatic morphology and function in a porcine model of endotoxaemia. Experimental study. 15 juvenile pigs. Flow probes were placed around the hepatic artery and portal vein. Catheters were introduced into the portal and hepatic veins, pulmonary artery, and aorta. Infusion of AE-ITU was started one hour before that of endotoxin (study group n = 6); thereafter both substances were infused simultaneously until the end of the study (6 hours). The controls (n = 9) had endotoxin alone. Hepatic morphology assessed by light and electron microscopy; and hepatic integrity and function by transaminase activities and oxygen consumption. Systemic, pulmonary, and hepatic blood flow and pressure. AE-ITU maintained systemic blood pressure (p < 0.05 compared with controls) without causing pulmonary hypertension. Neither hepatic morphology nor function were adversely influenced. In endotoxaemia AE-ITU has a favourable haemodynamic profile which is achieved without impairment of hepatic function or morphology.
Sigma-Aldrich
2-(2-Aminoethyl)isothiourea dihydrobromide