Merck
  • tBID, a membrane-targeted death ligand, oligomerizes BAK to release cytochrome c.
tBID, a membrane-targeted death ligand, oligomerizes BAK to release cytochrome c.
Genes & development (2000-08-19)
M C Wei, T Lindsten, V K Mootha, S Weiler, A Gross, M Ashiya, C B Thompson, S J Korsmeyer
ABSTRACT
TNFR1/Fas engagement results in the cleavage of cytosolic BID to truncated tBID, which translocates to mitochondria. Immunodepletion and gene disruption indicate BID is required for cytochrome c release. Surprisingly, the three-dimensional structure of this BH3 domain-only molecule revealed two hydrophobic alpha-helices suggesting tBID itself might be a pore-forming protein. Instead, we demonstrate that tBID functions as a membrane-targeted death ligand in which an intact BH3 domain is required for cytochrome c release, but not for targeting. Bak-deficient mitochondria and blocking antibodies reveal tBID binds to its mitochondrial partner BAK to release cytochrome c, a process independent of permeability transition. Activated tBID results in an allosteric activation of BAK, inducing its intramembranous oligomerization into a proposed pore for cytochrome c efflux, integrating the pathway from death receptors to cell demise.
Sigma-Aldrich
BID human, ≥95% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous solution
Sigma-Aldrich
BID, Caspase-8-cleaved from mouse, ≥95% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous solution