Merck
  • The polypeptide encoded by the cDNA for human cell surface antigen Fas can mediate apoptosis.
The polypeptide encoded by the cDNA for human cell surface antigen Fas can mediate apoptosis.
Cell (1991-07-26)
N Itoh, S Yonehara, A Ishii, M Yonehara, S Mizushima, M Sameshima, A Hase, Y Seto, S Nagata
ABSTRACT
Mouse anti-Fas monoclonal antibody has a cytolytic activity on human cells that express the antigen. Complementary DNAs encoding the cell surface antigen Fas were isolated from a cDNA library of human T cell lymphoma KT-3 cells. The nucleotide sequence of the cDNAs revealed that the molecule coding for the Fas antigen determinant is a 319 amino acid polypeptide (Mr 36,000) with a single transmembrane domain. The extracellular domain is rich in cysteine residue, and shows a similarity to that of human tumor necrosis factor receptors, human nerve growth factor receptor, and human B cell antigen CD40. Murine WR19L cells or L929 cells transformed with the human Fas antigen cDNA were killed by the anti-Fas antibody in the process known as apoptosis.
Sigma-Aldrich
FAS Receptor human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Fas/Fc Chimera from mouse, >97% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture