Merck
  • Electrochemical and spectrometric study of antioxidant activity of pomiferin, isopomiferin, osajin and catalposide.
Electrochemical and spectrometric study of antioxidant activity of pomiferin, isopomiferin, osajin and catalposide.
Journal of pharmaceutical and biomedical analysis (2008-07-04)
Vaclav Diopan, Petr Babula, Violetta Shestivska, Vojtech Adam, Milan Zemlicka, Margita Dvorska, Jaromir Hubalek, Libuse Trnkova, Ladislav Havel, Rene Kizek
ABSTRACT
The antioxidant properties of pomiferin, isopomiferin, osajin and catalposide are evaluated. The electrochemical behaviour of these compounds at a carbon paste electrode was studied using square wave voltammetry. Oxidative signals, optimized frequencies and appropriate pH acetate buffer conditions were determined. The detection limits (3S/N) for pomiferin, isopomiferin, osajin and catalposide were estimated to be 50 pg/ml, 800 pg/ml, 40 pg/ml and 10 ng/ml, respectively. Furthermore, spectrometric test was employed with 2,2-diphenyl-1-picrylhydrazyle (DPPH) to evaluate the antioxidant activities of these compounds. Based on the obtained results, the highest antioxidant activity measured by DPPH tests was found at pomiferin followed by isopomiferin. The activities of osajin and catalposide were undetectable. The protective effects of pomiferin, isopomiferin, osajin and catalposide on DNA exposed to oxygen radicals in vitro were also studied. Changes in height of oxidative signals for the four bases (guanine, thymine, adenine and cytosine) were measured for DNA exposed to oxygen radicals, generated by Fenton's reaction, non-oxidized ssDNA (50 microg/ml) displayed well developed signals; however, after oxidative damage the observed oxidative signals decreased. Significant protective effects were observed for pomiferin and osajin. Decreased effect was observed for isopomiferin while a further reduced protective effect was seen for DNA exposed to catalposide. Based on the obtained results, pomiferin had the highest antioxidant activity followed by isopomiferin, osajin and catalposide.
Sigma-Aldrich
Catalposide, ≥95% (LC/MS-ELSD)
Sigma-Aldrich
Osajin, ≥95% (LC/MS-ELSD)