Merck
  • Taurolidine lock: the key to prevention of recurrent catheter-related bloodstream infections.
Taurolidine lock: the key to prevention of recurrent catheter-related bloodstream infections.
Clinical nutrition (Edinburgh, Scotland) (2005-05-18)
Brian Jurewitsch, Khursheed N Jeejeebhoy
ABSTRACT
The literature shows that repeated courses of antibiotics and catheter removals in a subset of patients suffering from multiple catheter-related bloodstream infections (CRBSI), are unlikely to prevent recurrence. In acceding to preventative strategies, we report our application of the antimicrobial chemotherapeutic Taurolidine used as a daily flush solution in seven home TPN patients suffering from multiple episodes. A pretreatment infection rate of 10.8 infections per 1000 catheter days decreased to 0.8 after treatment.
Sigma-Aldrich
Taurolidine, >97% (NMR), powder