Merck
  • Radiation-induced non-targeted response in vivo: role of the TGFβ-TGFBR1-COX-2 signalling pathway.
Radiation-induced non-targeted response in vivo: role of the TGFβ-TGFBR1-COX-2 signalling pathway.
British journal of cancer (2013-02-16)
Y Chai, R K K Lam, G M Calaf, H Zhou, S Amundson, T K Hei
ABSTRACT
Previous studies from our group and others have shown that cyclooxygenase-2 (COX-2) has an essential role in radiation-induced non-targeted responses and genomic instability in vivo. However, the signalling pathways involved in such effects remain unclear. A 1 cm(2) area (1 cm × 1 cm) in the lower abdominal region of gpt delta transgenic mice was irradiated with 5 Gy of 300 keV X-rays. Nimesulide, a selective COX-2 inhibitor, was given to mice for five consecutive days before irradiation. Changes in transforming growth factor-beta (TGF-β) and TGF-β receptor type-1 (TGFBR1) mediated signalling pathways, in the out of radiation field lung and liver tissues were examined. While the plasma level of cytokines remained unchanged, the expression of TGF-β and its receptors was elevated in non-targeted lung tissues after partial body irradiation. In contrast to the predominant expression of TGF-β in stromal and alveolar cells, but not in bronchial epithelial cells, TGF-β receptors, especially TGFBR1 were significantly elevated in non-targeted bronchial epithelial cells, which is consistent with the induction of COX-2. The different expression levels of TGFBR1 between liver and lung resulted in a tissue specific induction of COX-2 in these two non-targeted tissues. Multiple TGF-β induced signalling pathways were activated in the non-targeted lung tissues. The TGFβ-TGFBR1-COX-2 Signalling Pathway has a critical role in radiation-induced non-targeted response in vivo.
Sigma-Aldrich
Fructose-6-phosphate Kinase from Bacillus stearothermophilus, Type VII, lyophilized powder, ≥50 units/mg protein
Nimesulide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Nimesulide