Merck
  • Notes from the field: hospital outbreak of carbapenem-resistant Klebsiella pneumoniae producing New Delhi metallo-beta-lactamase--Denver, Colorado, 2012.
Notes from the field: hospital outbreak of carbapenem-resistant Klebsiella pneumoniae producing New Delhi metallo-beta-lactamase--Denver, Colorado, 2012.
MMWR. Morbidity and mortality weekly report (2013-02-15)
ABSTRACT
On August 16, 2012, the Colorado Department of Public Health and Environment was notified of two patients at an acute-care hospital in Denver with carbapenem-resistant Enterobacteriaceae (CRE), specifically Klebsiella pneumoniae (CRKP), isolated from respiratory specimens during July-August. Both isolates produced New Delhi metallo-beta-lactamase (NDM). A review of microbiology records identified a third patient with NDM-producing CRKP isolated from a respiratory specimen, admitted in May. Active surveillance cultures in September identified an additional five patients colonized with NDM-producing CRKP. An investigation was launched by the hospital and the Colorado Department of Public Health and Environment to guide infection control measures and limit transmission.
Sigma-Aldrich
Penicillinase from Bacillus cereus, lyophilized, powder, white, ~13 U/mg
Sigma-Aldrich
Penicillinase from Bacillus cereus, lyophilized powder, 1,500-3,000 units/mg protein (using benzylpenicillin)
Sigma-Aldrich
β-Lactamase, recombinant, expressed in E. coli
Sigma-Aldrich
β-Lactamase from Enterobacter cloacae, Type IV, lyophilized powder, 0.2-0.6 units/mg protein (using benzylpenicillin)
Sigma-Aldrich
β-Lactamase from Pseudomonas fluorescens, lyophilized powder, recombinant, expressed in E. coli