- Horizontal gene transfer: you are what you eat.
Horizontal gene transfer: you are what you eat.
Biochemical and biophysical research communications (2010-05-25)
Lars Holmgren
PMID20494129
ABSTRACT
Horizontal or lateral gene transfer is an effective mechanism for the exchange of genetic information in bacteria allowing bacterial diversification and facilitating adaptation to new environments. Recent data demonstrate that DNA may also be transferred between somatic cells via the uptake of apoptotic bodies. This process allows transfer of viral genes that have been incorporated into the genome in a receptor-independent fashion. Transferred DNA is replicated and propagated in daughter cells in cell that have an inactivated DNA response which may impact tumor progression.
Sigma-Aldrich
Deoxyribonuclease II from porcine spleen, Type IV, lyophilized powder, 2,000-6,000 Kunitz units/mg protein (biuret)
Sigma-Aldrich
Deoxyribonuclease II from bovine spleen, Type V, essentially salt-free, lyophilized powder, ≥1,000 units/mg protein