Merck
  • Denitrification by the fungus Fusarium oxysporum involves NADH-nitrate reductase.
Denitrification by the fungus Fusarium oxysporum involves NADH-nitrate reductase.
Bioscience, biotechnology, and biochemistry (2008-02-08)
Tatsuya Fujii, Naoki Takaya
ABSTRACT
Fusarium oxysporum JCM11502 expresses a denitrifying (nitrate (NO(3)(-))-respiring) mechanism and can thrive under oxygen (O(2)) limitation. The fungus reduces NO(3)(-) to nitrite at the initial step of denitrification. In this study, we cloned the gene coding NADH-NO(3)(-) reductase (NADH-Nar) (niaD) from F. oxysporum JCM11502. The niaD gene complemented the defective NO(3)(-) assimilation by mutant strain M10, indicating that the fungus reduced NO(3)(-) through NADH-Nar activity and assimilated it like other fungi. We found that the transcription of niaD and the production of NADH-Nar activity were enhanced under O(2)-limited denitrifying conditions relative to aerobic conditions. Strain M10 produced less NADH-Nar activity and less denitrified product than the wild-type strain. Introducing niaD into the mutant also restored these defects, indicating that niaD is involved in denitrification. These results indicate that the fungus denitrified NO(3)(-) through NADH-Nar activity in addition to the ubiquinol-Nar mechanism.
Sigma-Aldrich
Nitrate Reductase from Arabidopsis thaliana, vial of ≥0.5 unit