Merck
  • Variation in aspects of cysteine proteinase catalytic mechanism deduced by spectroscopic observation of dithioester intermediates, kinetic analysis and molecular dynamics simulations.
Variation in aspects of cysteine proteinase catalytic mechanism deduced by spectroscopic observation of dithioester intermediates, kinetic analysis and molecular dynamics simulations.
The Biochemical journal (2001-07-06)
J D Reid, S Hussain, S K Sreedharan, T S Bailey, S Pinitglang, E W Thomas, C S Verma, K Brocklehurst
ABSTRACT
The possibility of a slow post-acylation conformational change during catalysis by cysteine proteinases was investigated by using a new chromogenic substrate, N-acetyl-Phe-Gly methyl thionoester, four natural variants (papain, caricain, actinidin and ficin), and stopped-flow spectral analysis to monitor the pre-steady state formation of the dithioacylenzyme intermediates and their steady state hydrolysis. The predicted reversibility of acylation was demonstrated kinetically for actinidin and ficin, but not for papain or caricain. This difference between actinidin and papain was investigated by modelling using QUANTA and CHARMM. The weaker binding of hydrophobic substrates, including the new thionoester, by actinidin than by papain may not be due to the well-known difference in their S2-subsites, whereby that of actinidin in the free enzyme is shorter due to the presence of Met211. Molecular dynamics simulation suggests that during substrate binding the sidechain of Met211 moves to allow full access of a Phe sidechain to the S2-subsite. The highly anionic surface of actinidin may contribute to the specificity difference between papain and actinidin. During subsequent molecular dynamics simulations the P1 product, methanol, diffuses rapidly (over<8 ps) out of papain and caricain but 'lingers' around the active centre of actinidin. Uniquely in actinidin, an Asp142-Lys145 salt bridge allows formation of a cavity which appears to constrain diffusion of the methanol away from the catalytic site. The cavity then undergoes large scale movements (over 4.8 A) in a highly correlated manner, thus controlling the motions of the methanol molecule. The changes in this cavity that release the methanol might be those deduced kinetically.
Sigma-Aldrich
Ficin from fig tree latex, powder, ≥0.1 unit/mg solid
Sigma-Aldrich
Ficin from fig tree latex, lyophilized powder
Sigma-Aldrich
Ficin from fig tree latex, saline suspension, ≥1.0 units/mg protein (biuret)