Merck
  • Synthesis, characterization, and biological activity studies of copper(II) mixed compound with histamine and nalidixic acid.
Synthesis, characterization, and biological activity studies of copper(II) mixed compound with histamine and nalidixic acid.
Bioinorganic chemistry and applications (2009-06-27)
Egla Yareth Bivián-Castro, Mercedes G López, Mario Pedraza-Reyes, Sylvain Bernès, Guillermo Mendoza-Díaz
ABSTRACT
A mixed copper complex with deprotonated nalidixic acid (nal) and histamine (hsm) was synthesized and characterized by FTIR, UV-Vis, elemental analysis, and conductivity. The crystal structure of [Cu(hsm)(nal)H(2)O]Cl.3H(2)O (chn) showed a pentacoordinated cooper(II) in a square pyramidal geometry surrounded by two N atoms from hsm, two O atoms from the quinolone, and one apical water oxygen. Alteration of bacterial DNA structure and/or associated functions in vivo by [Cu(hsm)(nal)H(2)O]Cl.3H(2)O was demonstrated by the induction of a recA-lacZ fusion integrated at the amyE locus of a recombinant Bacillus subtilis strain. Results from circular dichroism and denaturation of calf thymus DNA (CT-DNA) suggested that increased amounts of copper complex were able to stabilize the double helix of DNA in vitro mainly by formation of hydrogen bonds between chn and the sugars of DNA minor groove. In vivo and in vitro biological activities of the chn complex were compared with the chemical nuclease [Cu(phen)(nal)H(2)O]NO(3) . 3H(2)O (cpn) where phen is phenanthroline.
Sigma-Aldrich
Histamine dihydrochloride, ≥99% (TLC), powder
Sigma-Aldrich
Histamine dihydrochloride, ≥99.0% (AT)