Merck
Coronin 2B deficiency induces nucleolar stress and neuronal apoptosis.
Cell death & disease (2024-06-28)
Hongjiao Wu, Yujie Yang, Wanying Yi, Yue Qiu, Shuangshuang Ma, Jinying Xu, Yingying Fan, Yuewen Chen, Yu Chen
ABSTRACT
In eukaryotes, the nucleolus is the critical non-membranous organelle within nuclei that is responsible for ribosomal DNA (rDNA) transcription and ribosome biogenesis. The transcription of rDNA, a rate-limiting step for ribosome biogenesis, is tightly regulated to meet the demand for global protein synthesis in response to cell physiology, especially in neurons, which undergo rapid changes in morphology and protein composition during development and synaptic plasticity. However, it is unknown how the pre-initiation complex for rDNA transcription is efficiently assembled within the nucleolus in neurons. Here, we report that the nucleolar protein, coronin 2B, regulates rDNA transcription and maintains nucleolar function through direct interaction with upstream binding factor (UBF), an activator of RNA polymerase I transcriptional machinery. We show that coronin 2B knockdown impairs the formation of the transcription initiation complex, inhibits rDNA transcription, destroys nucleolar integrity, and ultimately induces nucleolar stress. In turn, coronin 2B-mediated nucleolar stress leads to p53 stabilization and activation, eventually resulting in neuronal apoptosis. Thus, we identified that coronin 2B coordinates with UBF to regulate rDNA transcription and maintain proper nucleolar function in neurons.
Sigma-Aldrich
Anti-CORO2B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-β-Tubulin III (neuronal) antibody, Mouse monoclonal, ~1.0 mg/mL, clone 2G10, purified from hybridoma cell culture
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture