Merck
  • Bacterial cupredoxin azurin hijacks cellular signaling networks: Protein-protein interactions and cancer therapy.
Bacterial cupredoxin azurin hijacks cellular signaling networks: Protein-protein interactions and cancer therapy.
Protein science : a publication of the Protein Society (2017-09-30)
Meng Gao, Jingjing Zhou, Zhengding Su, Yongqi Huang
ABSTRACT
Azurin secreted by Pseudomonas aeruginosa is an anticancer bacteriocin, which preferentially enters human cancer cells and induces apoptosis or growth inhibition. It turns out that azurin is a multi-target anticancer agent interfering in the p53 signaling pathway and the non-receptor tyrosine kinases signaling pathway. This suggests that azurin exerts its anticancer activity by interacting with multiple targets and interfering in multiple steps in disease progression. Therefore, azurin could overcome resistance to therapy. Besides azurin, putative bacteriocins that possess functional properties similar to those of azurin have been identified in more bacteria species. A systematic investigation on the anticancer mechanisms of azurin and the azurin-like bacteriocins will provide more and better options in cancer therapy. In this review, we summarize how azurin and the derived peptides hijack key cellular regulators or cell surface receptors to remodel the cellular signaling networks. In particular, we highlight the necessity of determining the structure of azurin/p53 complex and investigating the influence of post-translational modifications on interactions between azurin and p53. Therapeutic applications of azurin and derived peptides are also discussed.
Sigma-Aldrich
Azurin, from Pseudomonas aeruginosa, lyophilized powder