Merck
Dynamics of cell rounding during detachment.
iScience (2023-05-12)
Agata Nyga, Katarzyna Plak, Martin Kräter, Marta Urbanska, Kyoohyun Kim, Jochen Guck, Buzz Baum
ABSTRACT
Animal cells undergo repeated shape changes, for example by rounding up and respreading as they divide. Cell rounding can be also observed in interphase cells, for example when cancer cells switch from a mesenchymal to an ameboid mode of cell migration. Nevertheless, it remains unclear how interphase cells round up. In this article, we demonstrate that a partial loss of substrate adhesion triggers actomyosin-dependent cortical remodeling and ERM activation, which facilitates further adhesion loss causing cells to round. Although the path of rounding in this case superficially resembles mitotic rounding in involving ERM phosphorylation, retraction fiber formation, and cortical remodeling downstream of ROCK, it does not require Ect2. This work provides insights into the way partial loss of adhesion actives cortical remodeling to drive cell detachment from the substrate. This is important to consider when studying the mechanics of cells in suspension, for example using methods like real-time deformability cytometry (RT-DC).
Sigma-Aldrich
Calyculin A from Discodermia calyx, ≥90% (HPLC), solid
Sigma-Aldrich
Ezrin Inhibitor, NSC668394, The Ezrin Inhibitor, NSC668394 controls the biological activity of Ezrin.
Sigma-Aldrich
Latrunculin B from Latruncula magnifica, ≥80% (HPLC), solid
Sigma-Aldrich
Y-27632 dihydrochloride, ≥98% (HPLC)