Merck
  • Differential roles of type I interferon signaling in tumor versus host cells in experimental glioma models.
Differential roles of type I interferon signaling in tumor versus host cells in experimental glioma models.
Translational oncology (2022-12-27)
Evelina Blomberg, Manuela Silginer, Patrick Roth, Michael Weller
ABSTRACT
Despite multimodal treatment approaches including surgery, radiotherapy and chemotherapy, the median survival for patients with glioblastoma remains in the range of one year and thus poor. Type I interferons (IFN) are involved in immune responses to viral infection and exhibit anti-tumor activity in certain cancers. Here we explored the biological relevance of constitutive type I IFN signaling in murine glioma models in vitro and in vivo. CT-2A, GL-261, SMA-497, SMA-540 and SMA-560 murine glioma cells expressed IFN type I receptors IFNAR1 and IFNAR2 and were responsive to exogenous IFN stimulation. CRISPR/Cas9-mediated deletion of IFNAR1 decreased the baseline expression of type I IFN response genes in GL-261 cells, but neither in CT-2A nor in SMA-560 cells. IFNAR1 deletion slowed growth in GL-261 and SMA-560, but not in CT-2A cells. However, only the growth of IFNAR1-depleted GL-261 tumors and not that of SMA-560 tumors was delayed in vivo upon orthotopic tumor cell implantation into syngeneic mice. This survival gain was no longer detected when the IFNAR1-depleted GL-261 cells were inoculated into IFNAR1-deficient mice. Altogether these data suggest that constitutive type I IFN signaling in gliomas may be pro-tumorigenic, but only in a microenvironment that is proficient for type I IFN signaling in the host.
Sigma-Aldrich
CT-2A Mouse Glioma Cell Line, CT-2A mouse glioma cell line is a valuable mouse model for therapeutic research on brain malignancies.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution