Merck
  • RABENOSYN separation-of-function mutations uncouple endosomal recycling from lysosomal degradation, causing a distinct Mendelian disorder.
RABENOSYN separation-of-function mutations uncouple endosomal recycling from lysosomal degradation, causing a distinct Mendelian disorder.
Human molecular genetics (2022-06-03)
Franziska Paul, Calista Ng, Umar Bin Mohamad Sahari, Shahriar Nafissi, Yalda Nilipoor, Ali Reza Tavasoli, Carine Bonnard, Pui-Mun Wong, Nasrinsadat Nabavizadeh, Umut Altunoğlu, Mehrdad A Estiar, Charles B Majoie, Hane Lee, Stanley F Nelson, Ziv Gan-Or, Guy A Rouleau, Paul P Van Veldhoven, Rami Massie, Raoul C Hennekam, Ariana Kariminejad, Bruno Reversade
ABSTRACT
Rabenosyn (RBSN) is a conserved endosomal protein necessary for regulating internalized cargo. Here, we present clinical, genetic, cellular and biochemical evidence that two distinct RBSN missense variants are responsible for a novel Mendelian disorder consisting of progressive muscle weakness, facial dysmorphisms, ophthalmoplegia and intellectual disability. Using exome sequencing, we identified recessively acting germline alleles p.Arg180Gly and p.Gly183Arg, which are both situated in the FYVE domain of RBSN. We find that these variants abrogate binding to its cognate substrate phosphatidylinositol 3-phosphate (PI3P) and thus prevent its translocation to early endosomes. Although the endosomal recycling pathway was unaltered, mutant p.Gly183Arg patient fibroblasts show accumulation of cargo tagged for lysosomal degradation. Our results suggest that these variants are separation-of-function alleles, which cause a delay in endosomal maturation without affecting cargo recycling. We conclude that distinct germline mutations in RBSN cause non-overlapping phenotypes with specific and discrete endolysosomal cellular defects.
Sigma-Aldrich
Anti-RBSN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution