Merck
  • Functional expression of a rapidly inactivating neuronal calcium channel.
Functional expression of a rapidly inactivating neuronal calcium channel.
Nature (1993-06-03)
P T Ellinor, J F Zhang, A D Randall, M Zhou, T L Schwarz, R W Tsien, W A Horne
ABSTRACT
Diverse types of calcium channels in vertebrate neurons are important in linking electrical activity to transmitter release, gene expression and modulation of membrane excitability. Four classes of Ca2+ channels (T, N, L and P-type) have been distinguished on the basis of their electrophysiological and pharmacological properties. Most of the recently cloned Ca2+ channels fit within this functional classification. But one major branch of the Ca2+ channel gene family, including BII (ref. 15) and doe-1 (ref. 16), has not been functionally characterized. We report here the expression of doe-1 and show that it is a high-voltage-activated (HVA) Ca2+ channel that inactivates more rapidly than previously expressed calcium channels. Unlike L-type or P-type channels, doe-1 is not blocked by dihydropyridine antagonists or the peptide toxin omega-Aga-IVA, respectively. In contrast to a previously cloned N-type channel, doe-1 block by omega-CTx-GVIA requires micromolar toxin and is readily reversible. Unlike most HVA channels, doe-1 also shows unusual sensitivity to block by Ni2+. Thus, doe-1 is an HVA Ca2+ channel with novel functional properties. We have identified a Ca2+ channel current in rat cerebellar granule neurons that resembles doe-1 in many kinetic and pharmacological features.