Merck
  • Interaction among extenders, cryoprotectants and Orvus Es Paste supplementation and freezing rate for sperm cryopreservation in the dromedary camel.
Interaction among extenders, cryoprotectants and Orvus Es Paste supplementation and freezing rate for sperm cryopreservation in the dromedary camel.
Andrologia (2021-09-09)
Clara Malo, Ignacio De Blas, Julian Alexandra Skidmore
ABSTRACT
This study evaluated the effects of freezing extenders, cryoprotectants and their concentrations, presence of Orvus Es Paste and freezing rates for cryopreserving dromedary camel sperm. Semen (five males; 2 ejaculates/male) was frozen in one of the following extenders (Green Buffer® or INRA96® ), cryoprotectants (3 and 6% glycerol or ethylene glycol), with or without Orvus Es Paste and freezing at two different heights (1 and 4 cm) above liquid nitrogen. Sperm motility recovery parameters were evaluated post-thaw (0 and 1 hr), vitality and acrosome integrity (0 hr). Green Buffer showed higher total motility recoveries (p < .001). Higher percentage of cryoprotectant improved both total and progressive motility at 0 hr (p < .001; p = .003) and 1 hr (p < .001; p = .005). Acrosome integrity at thawing increased in the presence of ethylene glycol (p < .001) and Orvus Es Paste (p = .001). Kinematics were affected by extender, cryoprotectant concentration and Orvus Es Paste at 0 and 1 hr, and type of cryoprotectant only influenced them at 0h. Our findings showed strong interactions among type of cryoprotectant and concentration, and extender and Orvus Es Paste. Generally, combining Green Buffer, 3%-6% ethylene glycol or 6% glycerol without Orvus Es Paste, regardless of the freezing rates, yielded the highest post-thaw parameters for camel sperm.
Sigma-Aldrich
E-64, protease inhibitor
Sigma-Aldrich
Papain from papaya latex, lyophilized powder, aseptically filled