Merck
  • Early Stepdown Weaning of Dairy Calves with Glutamine and Branched-Chain Amino Acid Supplementations.
Early Stepdown Weaning of Dairy Calves with Glutamine and Branched-Chain Amino Acid Supplementations.
Animals : an open access journal from MDPI (2022-06-25)
Janaka Wickramasinghe, Can Ayhan Kaya, Donald Beitz, Ranga Appuhamy
ABSTRACT
The study objective was to examine the effects of supplementing Gln and BCAA on the SFI and ADG of weaning dairy calves. Holstein heifer calves (11 calves /treatment) at 35 d of age were assigned to: (1) no amino acids (CTL), (2) Gln (8.0 g/d) alone (GLN), or (3) Gln (8.0 g/d) and BCAA (GLNB; 17.0, 10.0, and 11.0 g/d leucine, isoleucine, and valine, respectively) supplementations in whole milk during a stepdown weaning scheme. Calves were weaned completely once they achieved ≥1.0 kg/d SFI. Neither GLN nor GLNB affected SFI or ADG in the first week during weaning. The GLNB decreased SFI compared to CTL, but the SFI was similar between CTL and GLN in the remainder of the weaning scheme. All calves were weaned at 50 d of age. The SFI of GLNB was lower than that of GLN, and the SFI of both GLN and GLNB were lower than CTL post-weaning. The decreased SFI did not alter ADG during weaning or post-weaning. The GLNB tended to have higher plasma leptin and lower plasma serotonin concentrations compared to CTL. Glutamine and BCAA seem to affect the SFI of calves by modulating the secretions of endocrine cells in the gastrointestinal tract.