Merck
  • MicroRNA miR-24-3p Mediates the Negative Regulation of Lipopolysaccharide-Induced Endometrial Inflammatory Response by Targeting TNF Receptor-Associated Factor 6 (TRAF6).
MicroRNA miR-24-3p Mediates the Negative Regulation of Lipopolysaccharide-Induced Endometrial Inflammatory Response by Targeting TNF Receptor-Associated Factor 6 (TRAF6).
Journal of inflammation research (2022-02-18)
Ayodele Olaolu Oladejo, Yajuan Li, Bereket Habte Imam, Xiaoyu Ma, Wenxiang Shen, Xiaohu Wu, Wei Jiang, Jie Yang, Yanan Lv, Xuezhi Ding, Shengyi Wang, Zuoting Yan
ABSTRACT
Endometritis is a female reproductive disease that affects the cattle industries development and microRNAs (miRNAs) play a pivotal role and critical regulators of the innate immune response in varieties of diseases. The present study intends to investigate the regulatory role of miR-24-3p in the innate immune response involved in endometritis and evaluate its therapeutic potential. Whole mice uteri and bovine endometrial epithelial cells (BEECs) were separately stimulated with LPS. The BEECs were also transfected with miR-24-3p mimic and negative control; siTRAF6 and siNC; pcDNA3.1 empty and pcDNA3.1(+)TRAF6 separately with LPS stimulation. The expression levels of miR-24-3p and TRAF6 were measured via quantitative real-time polymerase chain reaction (qRT-PCR) and Western blot, respectively. LPS-induced inflammatory response assessed by inflammatory cytokines secretion and expression via ELISA and qRT-PCR. Bioinformatics analysis and luciferase reporter assay validated the interaction between miR-24-3p and TRAF6. The activation of the NF-ĸB/MAPK pathway and p65 phosphorylation was investigated by Western blot and immunofluorescence assay, respectively. The expression of miR-24-3p was decreased, and TRAF6 was elevated with an increased level of pro-inflammatory cytokines in LPS-treated BEECs and mice uterus. The overexpression of miR-24-3p suppressed LPS-induced secretion of inflammatory cytokines (IL-1β, IL-6, IL-8 and TNF-α) and deactivation of NF-ĸB/MAPK pathways. The downregulation of TRAF6 inhibited LPS-induced inflammatory response in BEECs. TRAF6 is validated as a target of miR-24-3p, and miR-24-3p reversed the overexpression of cloned TRAF6 on inflammation response in BEECs. Our findings demonstrate that the overexpression of miR-24-3p attenuates endometrial inflammation and the expression of pro-inflammatory mediators via suppressing TRAF6. Therefore, modulating the pathogenesis of endometritis and possibly, a therapeutic potential against endometritis.
Sigma-Aldrich
Collagen Type I−FITC Conjugate from bovine skin, ~1 mg/mL, solution
Pricing and availability is not currently available.