- Determination of total folates in infant formula and adult nutritionals by trienzyme extraction and UPLC-MS/MS Quantitation: First Action 2011.06.
Determination of total folates in infant formula and adult nutritionals by trienzyme extraction and UPLC-MS/MS Quantitation: First Action 2011.06.
The method for "Determination of Total Folates in Infant Formula and Adult Nutritionals by Trienzyme Extraction and UPLC-MS/MS Quantitation" was submitted to the Folate Working Group for consideration for adoption as Official First Action by AOAC INTERNATIONAL. This method uses ultra performance liquid chromatography/tandem mass spectrometry (UPLC-MS/MS) to determine the total folates in infant formulas and adult nutritionals after trienzyme digestion. Deconjugation of the various folate polyglutamates to folate monoglutamates is achieved by using rat plasma conjugase after the sample digestion with protease and a-amylase during the trienzyme digestion process. This method shows linearity of folate concentrations in the range of 10-19 100 microg/100 g. Extension of the range to cover folate concentrations of 5-2 000 000 microg/100 g can be achieved with appropriate adjustment of the sample weight and SPE cleanup loading volume. The recoveries ranged from 94.10 to 101.34%.