Merck
  • Evaluation of sperm quality of Erythrolamprus poecilogyrus sublineatus (Cope, 1860) (Serpentes, Dipsadidae).
Evaluation of sperm quality of Erythrolamprus poecilogyrus sublineatus (Cope, 1860) (Serpentes, Dipsadidae).
Brazilian journal of biology = Revista brasleira de biologia (2017-01-19)
A C Silva, A S Varela, T F Cardoso, E F Silva, D Loebmann, C D Corcini
ABSTRACT
Erythrolamprus poecilogyrus sublineatus (Cope, 1860), is a species widely distributed in the Pampa Domain, occurring in Rio Grande do Sul, Argentina and Uruguay, mainlyin the pampa region. In the coastal region of southern Brazil this is serpent is considered one of the most abundant. The purpose of the present study is to describe the techniques of sperm evaluation in vitro for E. poecilogyrus sublineatus in the coastal plain of Rio Grande do Sul, Brazil. After laparatomy the efferent vases were collected and the semen was diluted in 1ml Beltsville Thawing Solution. The characteristics of motility, membrane integrity, mitochondria, acrosome, DNA, cell viability and cellular functionality were evaluated. Fluorescent probes were used for the evaluation of sperm structure in epifluorescence microscope. With the techniques described, it was possible to identify intact and injured cells, enabling the determination of cell characteristics for the spring season (October and November). It was observed in the analyses that 80% of sperm cells were mobile and that 84.1 ± 8.0% of sperm membranes were intact. The standards found were of 48 ± 13.8% of intact acrosome, 73.6 ± 6.0 of perfect DNA and of 91.8 ± 4.0 of functional mitochondria. Thus, these values from the sperm analysis can be used as standards for the species Erythrolamprus poecilogyrus sublineatus.
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Sodium citrate dihydrate, ≥99%, FG