Merck
  • Niche-Mediated Integrin Signaling Supports Steady-State Hematopoiesis in the Spleen.
Niche-Mediated Integrin Signaling Supports Steady-State Hematopoiesis in the Spleen.
Journal of immunology (Baltimore, Md. : 1950) (2021-02-28)
Shubham Haribhau Mehatre, Irene Mariam Roy, Atreyi Biswas, Devila Prit, Sarah Schouteden, Joerg Huelsken, Catherine M Verfaillie, Satish Khurana
ABSTRACT
Outside-in integrin signaling regulates cell fate decisions in a variety of cell types, including hematopoietic stem cells (HSCs). Our earlier published studies showed that interruption of periostin (POSTN) and integrin-αv (ITGAV) interaction induces faster proliferation in HSCs with developmental stage-dependent functional effects. In this study, we examined the role of POSTN-ITGAV axis in lymphohematopoietic activity in spleen that hosts a rare population of HSCs, the functional regulation of which is not clearly known. Vav-iCre-mediated deletion of Itgav in the hematopoietic system led to higher proliferation rates, resulting in increased frequency of primitive HSCs in the adult spleen. However, in vitro CFU-C assays demonstrated a poorer differentiation potential following Itgav deletion. This also led to a decrease in the white pulp area with a significant decline in the B cell numbers. Systemic deletion of its ligand, POSTN, phenocopied the effects noted in Vav-Itgav-/- mice. Histological examination of Postn-deficient spleen also showed an increase in the spleen trabecular areas. Importantly, these are the myofibroblasts of the trabecular and capsular areas that expressed high levels of POSTN within the spleen tissue. In addition, vascular smooth muscle cells also expressed POSTN. Through CFU-S12 assays, we showed that hematopoietic support potential of stroma in Postn-deficient splenic hematopoietic niche was defective. Overall, we demonstrate that POSTN-ITGAV interaction plays an important role in spleen lymphohematopoiesis.
Sigma-Aldrich
Harris Hematoxylin Solution, Modified
Sigma-Aldrich
Eosin Y Solution, Alcoholic