Merck
  • Janibacter hoylei sp. nov., Bacillus isronensis sp. nov. and Bacillus aryabhattai sp. nov., isolated from cryotubes used for collecting air from the upper atmosphere.
Janibacter hoylei sp. nov., Bacillus isronensis sp. nov. and Bacillus aryabhattai sp. nov., isolated from cryotubes used for collecting air from the upper atmosphere.
International journal of systematic and evolutionary microbiology (2009-08-01)
S Shivaji, Preeti Chaturvedi, Zareena Begum, Pavan Kumar Pindi, R Manorama, D Ananth Padmanaban, Yogesh S Shouche, Shrikant Pawar, Parag Vaishampayan, C B S Dutt, G N Datta, R K Manchanda, U R Rao, P M Bhargava, J V Narlikar
ABSTRACT
Three novel bacterial strains, PVAS-1(T), B3W22(T) and B8W22(T), were isolated from cryotubes used to collect air samples at altitudes of between 27 and 41 km. Based on phenotypic characteristics, chemotaxonomic features, DNA-DNA hybridization with the nearest phylogenetic neighbours and phylogenetic analysis based on partial 16S rRNA gene sequences (PVAS-1(T), 1196 nt; B3W22(T), 1541 nt; B8W22(T), 1533 nt), the three strains were identified as representing novel species, and the names proposed are Janibacter hoylei sp. nov. (type strain PVAS-1(T) =MTCC 8307(T) =DSM 21601(T) =CCUG 56714(T)), Bacillus isronensis sp. nov. (type strain B3W22(T) =MTCC 7902(T) =JCM 13838(T)) and Bacillus aryabhattai sp. nov. (type strain B8W22(T) =MTCC 7755(T) =JCM 13839(T)).
Sigma-Aldrich
4-Ethylphenol, ≥97.0% (GC)