Merck
  • Adsorption of doxorubicin on poly(methyl methacrylate)-chitosan-heparin-coated activated carbon beads.
Adsorption of doxorubicin on poly(methyl methacrylate)-chitosan-heparin-coated activated carbon beads.
Langmuir : the ACS journal of surfaces and colloids (2012-02-09)
Jianjun Miao, Fuming Zhang, Majde Takieddin, Shaker Mousa, Robert J Linhardt
ABSTRACT
Extracorporeal filter cartridges, filled with an activated carbon bead (ACB) adsorbent, have been used for removal of overdosed cancer drugs from the blood. Coatings on adsorbent matrices, poly(methyl methacrylate) (PMMA)/activated carbon bead and PMMA/chitosan/heparin/ACB composites, were tested to improve their biocompatibility and blood compatibility. PMMA coating on ACBs was accomplished in a straightforward manner using a PMMA solution in ethyl acetate. A one-step hybrid coating of ACBs with PMMA-anticoagulant heparin required the use of acetone and water co-solvents. Multilayer coatings with three components, PMMA, chitosan, and heparin, involved three steps: PMMA was first coated on ACBs; chitosan was then coated on the PMMA-coated surface; and finally, heparin was covalently attached to the chitosan coating. Surface morphologies were studied by scanning electron microscopy. X-ray photoelectron spectroscopy confirmed the -SO(3)(-) group. Adsorption, of a chemotherapy drug (doxorubicin) from both water and phosphate-buffered saline, by the coated ACBs was examined. The adsorption isotherm curves were fitted using the Freundlich model. The current adsorption system might find potential applications in the removal of high-dose regional chemotherapy drugs while maintaining high efficiency, biocompatibility, and blood compatibility.
Sigma-Aldrich
Chitosan, high molecular weight