Merck
  • Femtosecond laser-assisted cataract surgery in shallow anterior chamber cases.
Femtosecond laser-assisted cataract surgery in shallow anterior chamber cases.
International ophthalmology (2020-10-26)
Andrés Fernández-Vega Cueto, Ignacio Rodríguez-Una, Pedro P Rodríguez-Calvo, José F Alfonso
ABSTRACT
To assess the effectiveness, safety and predictability of femtosecond laser-assisted cataract surgery (FLACS) in eyes with shallow anterior chamber (AC). This is a prospective consecutive clinical study. All eyes presented an anterior chamber depth (ACD) ≤ 2.1 mm and were submitted to FLACS with monofocal intraocular lens (mIOL) or trifocal intraocular lens (tIOL) implantation. Uncorrected distance visual acuity (VA) and corrected distance VA values were used to assess efficacy and safety of the surgery. Refraction, intraocular pressure (IOP), endothelial cell density (ECD) and ACD were evaluated before and 6 months post-surgery. Phacoemulsification was carried out successfully in all eyes, without intra- or postoperative complications. Efficacy and safety indexes were 0.96 and 1.26 for the mIOL group, and 0.87 and 1.01 for the tIOL group, respectively. The mean postoperative spherical equivalent was - 0.06 ± 0.28D and - 0.14 ± 0.38D for the mIOL and tIOL groups, respectively. The ECD varied from 2470 ± 483 to 2009 ± 538 cells/mm2 (p < 0.05) and from 2443 ± 319 to 2245 ± 628 cells/mm2 (p = 0.06) for the mIOL and tIOL groups, respectively. IOP significantly decreased (p < 0.05) after the surgery from 14.34 to 12.85 mmHg for the mIOL group and from 14.37 to 11.91 mmHg for the tIOL group, with a general reduction of medical hypotensive treatment in both groups (85% of cases required ≤ number of medications). ACD changed significantly (p < 0.05) from 1.96 ± 0.15 mm to 3.75 ± 0.30 mm after the surgery in the mIOL group and from 1.94 ± 0.15 mm to 3.23 ± 0.21 mm for the tIOL group. FLACS with implantation of either mIOL or tIOL may provide good efficacy, safety and predictability in eyes with shallow AC.