Merck
  • RNA m6 A methylation regulates uveal melanoma cell proliferation, migration, and invasion by targeting c-Met.
RNA m6 A methylation regulates uveal melanoma cell proliferation, migration, and invasion by targeting c-Met.
Journal of cellular physiology (2020-02-06)
Guangying Luo, Weiwei Xu, Yunping Zhao, Shanshan Jin, Siqi Wang, Qi Liu, Xiaoyan Chen, Jiao Wang, Feng Dong, Dan-Ning Hu, Peter S Reinach, Dongsheng Yan
ABSTRACT
N6 -methyladenosine (m6 A) is a novel epitranscriptomic marker that contributes to regulating diverse biological processes through controlling messenger RNA metabolism. However, it is unknown if m6 A RNA methylation affects uveal melanoma (UM) development. To address this question, we probed its function and molecular mechanism in UM. Initially, we demonstrated that global RNA m6 A methylation levels were dramatically elevated in both UM cell lines and clinical specimens. Meanwhile, we found that METTL3, a main m6 A regulatory enzyme, was significantly increased in UM cells and specimens. Subsequently, cycloleucine (Cyc) or METTL3 targeted small interfering RNA was used to block m6 A methylation in UM cells. We found that Cyc or silencing METTL3 significantly suppressed UM cell proliferation and colony formation through cell cycle G1 arrest, as well as migration and invasion by functional analysis. On the other hand, overexpression of METTL3 had the opposite effects. Furthermore, bioinformatics and methylated RNA immunoprecipitation-quantitative polymerase chain reaction identified c-Met as a direct target of m6 A methylation in UM cells. In addition, western blot analysis showed that Cyc or knockdown of METTL3 downregulated c-Met, p-Akt, and cell cycle-related protein levels in UM cells. Taken together, our results demonstrate that METTL3-mediated m6 A RNA methylation modulates UM cell proliferation, migration, and invasion by targeting c-Met. Such a modification acts as a critical oncogenic regulator in UM development.
Sigma-Aldrich
Cycloleucine, 97%