Merck
  • Celecoxib analogs possessing a N-(4-nitrooxybutyl)piperidin-4-yl or N-(4-nitrooxybutyl)-1,2,3,6-tetrahydropyridin-4-yl nitric oxide donor moiety: synthesis, biological evaluation and nitric oxide release studies.
Celecoxib analogs possessing a N-(4-nitrooxybutyl)piperidin-4-yl or N-(4-nitrooxybutyl)-1,2,3,6-tetrahydropyridin-4-yl nitric oxide donor moiety: synthesis, biological evaluation and nitric oxide release studies.
Bioorganic & medicinal chemistry letters (2010-01-26)
Morshed A Chowdhury, Khaled R A Abdellatif, Ying Dong, Gang Yu, Zhangjian Huang, Moshfiqur Rahman, Dipankar Das, Carlos A Velázquez, Mavanur R Suresh, Edward E Knaus
ABSTRACT
A new group of hybrid nitric oxide (NO) releasing anti-inflammatory (AI) coxib prodrugs (NO-coxibs) wherein the para-tolyl moiety present in celecoxib was replaced by a N-(4-nitrooxybutyl)piperidyl 15a-b, or N-(4-nitrooxybutyl)-1,2,3,6-tetrahydropyridyl 17a-b, NO-donor moiety was synthesized. All compounds released a low amount of NO upon incubation with phosphate buffered saline (PBS) at pH 7.4 (2.4-5.8% range). In comparison, the percentage NO released was higher (3.1-8.4% range) when these nitrate prodrugs were incubated in the presence of L-cysteine. In vitro COX-1/COX-2 isozyme inhibition studies showed this group of compounds are moderately more potent, and hence selective, inhibitors of the COX-2 relative to the COX-1 enzyme. AI structure-activity relationship data acquired showed that compounds having a MeSO2 COX-2 pharmacophore exhibited superior AI activity compared to analogs having a H2NSO2 substituent. Compounds having a MeSO2 COX-2 pharmacophore in conjunction with a N-(4-nitrooxybutyl)piperidyl (ED50=132.4 mg/kg po), or a N-(4-nitrooxybutyl)-1,2,3,6-tetrahydropyridyl (ED50=118.4 mg/kg po), moiety exhibited an AI potency profile that is similar to aspirin (ED50=128.7 mg/kg po) but lower than ibuprofen (ED50=67.4 mg/kg po).
Product Description
Supelco
Ibuprofen
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Aspirin, meets USP testing specifications