Merck
  • Metal-Organic Decomposition-Mediated Nanoparticulate Vanadium Oxide Hole Transporting Buffer Layer for Polymer Bulk-Heterojunction Solar Cells.
Metal-Organic Decomposition-Mediated Nanoparticulate Vanadium Oxide Hole Transporting Buffer Layer for Polymer Bulk-Heterojunction Solar Cells.
Polymers (2020-08-14)
Chengkai Xia, Won Tae Hong, Young Eun Kim, Woo-Seok Choe, Dong-Hwan Kim, Jung Kyu Kim
ABSTRACT
In this study, a solution-processable compact vanadium oxide (V2O5) film with a globular nanoparticulate structure is introduced to the hole transport layer (HTL) of polymer bulk-heterojunction based solar cells comprised of PTB7:PC70BM by using a facile metal-organic decomposition method to replace the conventionally utilized poly(3,4-ethylenedioxythiophene): poly(styrenesulfonate) (PEDOT:PSS). For this, a biocompatible structure-determining agent, polyethylene glycol (PEG, Mn 300), is used as an additive in the precursor to form the nanoparticulate compact V2O5 (hereafter referred to as NP-V2O5) film, which possesses an outstandingly smooth surface morphology. The introduction of NP-V2O5 HTL via the solution process with a neutral pH condition successfully improved the stability by preventing the decomposition of indium tin oxide (ITO) glass and the penetration of heavy-metal components and moisture, which are considered as the crucial drawbacks of using PEDOT:PSS. Over 1440 h (60 days) of the stability test, an organic solar cell (OSC) with NP-V2O5 showed a significant durability, maintaining 82% of its initial power conversion efficiency (PCE), whereas an OSC with PEDOT:PSS maintained 51% of its initial PCE. Furthermore, due to the positive effects of the modified surface properties of NP-V2O5, the PCE was slightly enhanced from 7.47% to 7.89% with a significant improvement in the short-circuit current density and fill factor.
Sigma-Aldrich
1,8-Diiodooctane, 98%, contains copper as stabilizer
Pricing and availability is not currently available.