Merck
  • AZD4320, A Dual Inhibitor of Bcl-2 and Bcl-xL, Induces Tumor Regression in Hematologic Cancer Models without Dose-limiting Thrombocytopenia.
AZD4320, A Dual Inhibitor of Bcl-2 and Bcl-xL, Induces Tumor Regression in Hematologic Cancer Models without Dose-limiting Thrombocytopenia.
Clinical cancer research : an official journal of the American Association for Cancer Research (2020-09-30)
Srividya B Balachander, Steven W Criscione, Kate F Byth, Justin Cidado, Ammar Adam, Paula Lewis, Terry Macintyre, Shenghua Wen, Deborah Lawson, Kathleen Burke, Tristan Lubinski, Jeffrey W Tyner, Stephen E Kurtz, Shannon K McWeeney, Jeffrey Varnes, R Bruce Diebold, Thomas Gero, Stephanos Ioannidis, Edward J Hennessy, William McCoull, Jamal C Saeh, Areya Tabatabai, Omid Tavana, Nancy Su, Alwin Schuller, Mathew J Garnett, Patricia Jaaks, Elizabeth A Coker, Gareth P Gregory, Andrea Newbold, Ricky W Johnstone, Eric Gangl, Martin Wild, Michael Zinda, J Paul Secrist, Barry R Davies, Stephen E Fawell, Francis D Gibbons
ABSTRACT
Targeting Bcl-2 family members upregulated in multiple cancers has emerged as an important area of cancer therapeutics. While venetoclax, a Bcl-2-selective inhibitor, has had success in the clinic, another family member, Bcl-xL, has also emerged as an important target and as a mechanism of resistance. Therefore, we developed a dual Bcl-2/Bcl-xL inhibitor that broadens the therapeutic activity while minimizing Bcl-xL-mediated thrombocytopenia. We used structure-based chemistry to design a small-molecule inhibitor of Bcl-2 and Bcl-xL and assessed the activity against in vitro cell lines, patient samples, and in vivo models. We applied pharmacokinetic/pharmacodynamic (PK/PD) modeling to integrate our understanding of on-target activity of the dual inhibitor in tumors and platelets across dose levels and over time. We discovered AZD4320, which has nanomolar affinity for Bcl-2 and Bcl-xL, and mechanistically drives cell death through the mitochondrial apoptotic pathway. AZD4320 demonstrates activity in both Bcl-2- and Bcl-xL-dependent hematologic cancer cell lines and enhanced activity in acute myeloid leukemia (AML) patient samples compared with the Bcl-2-selective agent venetoclax. A single intravenous bolus dose of AZD4320 induces tumor regression with transient thrombocytopenia, which recovers in less than a week, suggesting a clinical weekly schedule would enable targeting of Bcl-2/Bcl-xL-dependent tumors without incurring dose-limiting thrombocytopenia. AZD4320 demonstrates monotherapy activity in patient-derived AML and venetoclax-resistant xenograft models. AZD4320 is a potent molecule with manageable thrombocytopenia risk to explore the utility of a dual Bcl-2/Bcl-xL inhibitor across a broad range of tumor types with dysregulation of Bcl-2 prosurvival proteins.
Sigma-Aldrich
Tetramethylrhodamine ethyl ester perchlorate, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
SW48 Cells BAX/BAK (-/-,-/-)
Sigma-Aldrich
Boron-11B, 95 atom % 11B