Merck
  • CD44 engagement enhances acute myeloid leukemia cell adhesion to the bone marrow microenvironment by increasing VLA-4 avidity.
CD44 engagement enhances acute myeloid leukemia cell adhesion to the bone marrow microenvironment by increasing VLA-4 avidity.
Haematologica (2020-07-04)
Julia C Gutjahr, Elisabeth Bayer, Xiaobing Yu, Julia M Laufer, Jan P Höpner, Suzana Tesanovic, Andrea Härzschel, Georg Auer, Tanja Rieß, Astrid Salmhofer, Eva Szenes, Theresa Haslauer, Valerie Durand-Onayli, Andrea Ramspacher, Sandra P Pennisi, Marc Artinger, Nadja Zaborsky, Alexandre Chigaev, Fritz Aberger, Daniel Neureiter, Lisa Pleyer, Daniel F Legler, Veronique Orian-Rousseau, Richard Greil, Tanja N Hartmann
ABSTRACT
Adhesive properties of leukemia cells shape the degree of organ infiltration and the extent of leukocytosis. CD44 and the integrin VLA-4, a CD49d/CD29 heterodimer, are important factors of progenitor cell adhesion in bone marrow (BM). Here, we report their cooperation in acute myeloid leukemia (AML) by a novel non-classical CD44-mediated way of inside-out VLA-4 activation. In primary AML BM samples from patients and the OCI-AML3 cell line, CD44 engagement by hyaluronan induced inside-out activation of VLA-4 resulting in enhanced leukemia cell adhesion on VCAM-1. This was independent from VLA-4 affinity regulation but based on ligand-induced integrin clustering on the cell surface. CD44-induced VLA-4 activation could be inhibited by the Src family kinase inhibitor PP2 and the multikinase inhibitor midostaurin. In further consequence, the increased adhesion on VCAM-1 allowed AML cells to strongly bind stromal cells. Thereby VLA-4/VCAM-1 interaction promoted activation of Akt, MAPK, NF-kB and mTOR signaling and decreased AML cell apoptosis. Collectively, our investigations provide a mechanistic description of an unusual CD44 function in regulating VLA-4 avidity in AML, supporting AML cell retention in the supportive BM microenvironment.
Sigma-Aldrich
Anti-Integrin α4 Antibody, clone PS/2, clone PS/2, Chemicon®, from rat
Sigma-Aldrich
Anti-Integrin α4 Antibody, clone HP2/1, clone HP2/1, Chemicon®, from mouse