Merck
  • Anti-depressant-like effect of vitexin in BALB/c mice and evidence for the involvement of monoaminergic mechanisms.
Anti-depressant-like effect of vitexin in BALB/c mice and evidence for the involvement of monoaminergic mechanisms.
European journal of pharmacology (2012-10-27)
Özgür Devrim Can, Ümide Demir Özkay, Umut İrfan Üçel
ABSTRACT
The present study was designed to investigate the putative effect of vitexin, a flavone C-glucoside present in some drugs, medicinal plants and nutraceuticals, on the central nervous system. Vitexin (10-30 mg/kg) did not show significant alterations in the behaviour of mice tested in hole-board, plus-maze or activity cage tests. However, immobility time of the mice significantly reduced by vitexin administrations in both the tail-suspension and modified forced swimming tests. The anti-immobility effect of vitexin in the tail-suspension test was reversed with α-methyl-para-tyrosine methyl ester (AMPT, an inhibitor of catecholamine synthesis, 100mg/kg, i.p.), yohimbine (an α(2)-adrenoceptor antagonist, 1mg/kg, i.p.), NAN 190 (a 5-HT(1A) antagonist, 0.5mg/kg, i.p.), SCH 23390 (a dopamine D(1) antagonist, 0.05 mg/kg, s.c.) and sulpiride (a dopamine D(2)/D(3) antagonist, 50mg/kg, i.p.). The same effect was not reversed, however, by p-chlorophenylalanine methyl ester (PCPA; an inhibitor of serotonin synthesis 100mg/kg, i.p., administered for 4 consecutive days), ketanserin (a 5-HT(2A/2C) antagonist, 1-4 mg/kg, i.p.), ondansetron (a 5-HT(3) antagonist, 0.1-0.4 mg/kg, i.p.), prazosin (an α(1)-adrenoceptor antagonist, 1-4 mg/kg, i.p.), or propranolol (a non-selective β-adrenoceptor antagonist, 5-20mg/kg, i.p.). These results suggest that the anti-depressant-like effect of vitexin is mediated through an increase in catecholamine levels in the synaptic cleft as well as through interactions with the serotonergic 5-HT(1A), noradrenergic α(2), and dopaminergic D(1), D(2), and D(3) receptors. To our knowledge, this is the first study to show findings that indicate an anti-depressant-like effect of vitexin and its underlying mechanisms.
Sigma-Aldrich
Reboxetine mesylate hydrate, >96% (HPLC), solid