Merck
  • IRF2 regulates cellular survival and Lenvatinib-sensitivity of hepatocellular carcinoma (HCC) through regulating β-catenin.
IRF2 regulates cellular survival and Lenvatinib-sensitivity of hepatocellular carcinoma (HCC) through regulating β-catenin.
Translational oncology (2021-03-19)
Yarong Guo, Jun Xu, Qiang Du, Yihe Yan, David A Geller
ABSTRACT
Hepatocellular carcinoma (HCC) is the third leading cause of cancer-related death worldwide. Lenvatinib oral chemotherapy is approved as a first-line treatment of patients with unresectable HCC. The efficacy and therapeutic duration of lenvatinib are limited by drug resistance, and the mechanism is unclear. IRF2 is a constitutive transcription factor associated with the development of various cancers by regulating cancer cell growth, apoptosis, and drug resistance. However, the potential role of IRF2 in lenvatinib resistance in HCC has not been explored. In this study, we found that IRF2 promoted proliferation, inhibited apoptosis, and increased lenvatinib resistance of HCC cells by regulating β-catenin expression. Silencing IRF2 downregulated the expression of β-catenin, while overexpressing IRF2 upregulated β-catenin. Moreover, the expression of β-catenin and IRF2 was positively correlated in HCC tissues. Inhibiting β-catenin with XAV-939 effectively abrogated β-catenin expression caused by lenvatinib treatment. These findings identify an important function of IRF2 in HCC and demonstrate a mechanism of lenvatinib resistance of HCC cells. Targeting IRF2 may be a potential strategy to improve the therapeutic effect of lenvatinib on HCC.
Sigma-Aldrich
Monoclonal Anti-β-Catenin antibody produced in mouse, clone 15B8, ascites fluid