Merck
  • The miR-93-3p/ZFP36L1/ZFX axis regulates keratinocyte proliferation and migration during skin wound healing.
The miR-93-3p/ZFP36L1/ZFX axis regulates keratinocyte proliferation and migration during skin wound healing.
Molecular therapy. Nucleic acids (2021-01-22)
Xiao Feng, Shuangbai Zhou, Weilin Cai, Jincai Guo
ABSTRACT
Keratinocyte proliferation and migration are crucial steps during skin wound healing. The functional role of microRNAs (miRs) remains relatively unknown during this process. miR-93 levels have been reported to increase within 24 h of skin wound healing; however, whether miR-93-3p or miR-93-5p plays a specific role in wound healing is yet to be studied. In this study, with the use of an in vivo mouse skin wound-healing model, we demonstrate that miR-93-3p is significantly upregulated, whereas there is no change in the expression of miR-93-5p during skin wound healing. In HaCaT cells, miR-93-3p overexpression increased proliferation and migration of the cells, whereas miR-93-3p inhibition had the reverse effect. Additionally, it was evident that ZFP36L1 was a direct target of miR-93-3p in keratinocytes. Further, ZFP36L1 silencing mirrored the consequences observed during miR-93-3p overexpression on both proliferation and migration of keratinocytes. In addition, we demonstrate that zinc-finger X-linked (ZFX), as a target for ZFP36L1, is involved in the promotion of the miR-93-3p/ZFP36L1 axis in keratinocyte proliferation and migration. Ultimately, we found that mouse skin wound model treatment with anti-miR-93-3p delayed wound healing. Overall, our results show that miR-93-3p is a crucial regulator of skin wound healing that facilitates keratinocyte proliferation and migration through ZFP36L1/ZFX axis.
Sigma-Aldrich
Anti-ZFX, (N-terminal) antibody produced in rabbit, affinity isolated antibody