Merck
  • A simple and highly efficient method for multi-allelic CRISPR-Cas9 editing in primary cell cultures.
A simple and highly efficient method for multi-allelic CRISPR-Cas9 editing in primary cell cultures.
Cancer reports (Hoboken, N.J.) (2020-07-29)
Pia Hoellerbauer, Megan Kufeld, Sonali Arora, Hua-Jun Wu, Heather M Feldman, Patrick J Paddison
ABSTRACT
CRISPR-Cas9-based technologies have revolutionized experimental manipulation of mammalian genomes. None-the-less, limitations of the delivery and efficacy of these technologies restrict their application in primary cells. To create an optimized protocol for penetrant, reproducible, and fast targeted genome editing in cell cultures derived from primary cells, using patient-derived glioblastoma stem-like cells (GSCs) and human neural stem/progenitor cells (NSCs) for proof-of-concept experiments. We employed transient nucleofection of Cas9:sgRNA ribonucleoprotein complexes composed of chemically synthesized 2'-O-methyl 3'phosphorothioate-modified sgRNAs and purified Cas9 protein. Insertion-deletion mutation (indel) frequency and size distribution were measured via computational deconvolution of Sanger sequencing trace data. We found that this optimized technique routinely allows for >90% indel formation in only 3 days, without the need to create clonal lines for simple loss-of-function experiments. Using Western blotting, we observed near-total protein loss of target genes in cell pools. Additionally, we found that this approach allows for the creation of targeted genomic deletions. Furthermore, by using RNA-seq in edited NSCs to assess gene expression changes resulting from knockout of tumor suppressors commonly altered in glioblastoma, we also demonstrated the utility of this method for quickly creating a series of gene knockouts that allow for the study of oncogenic activities. Our data suggest that this relatively simple method can be used for highly efficient and fast gene knockout, as well as for targeted genomic deletions, even in hyperdiploid cells (such as GSCs). This represents an extremely useful tool for the cancer research community when wishing to inactivate not only coding genes, but also non-coding RNAs, UTRs, enhancers, and promoters. This method can be readily applied to diverse cell types by varying the nucleofection conditions.
Sigma-Aldrich
Anti-GAPDH, C-Terminal antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid