Merck
  • USP7-dependent control of myogenin stability is required for terminal differentiation in skeletal muscle progenitors.
USP7-dependent control of myogenin stability is required for terminal differentiation in skeletal muscle progenitors.
The FEBS journal (2020-03-03)
Eduardo de la Vega, Natalia González, Felipe Cabezas, Fabián Montecino, Natasha Blanco, Hugo Olguín
ABSTRACT
Satellite cells (SCs) are myogenic progenitors responsible for skeletal muscle regeneration and maintenance. Upon activation, SCs enter a phase of robust proliferation followed by terminal differentiation. Underlying this myogenic progression, the sequential expression of muscle regulatory transcription factors (MRFs) and the downregulation of transcription factor paired box gene 7 (Pax7) are key steps regulating SC fate. In addition to transcriptional regulation, post-translational control of Pax7 and the MRFs provides another layer of spatiotemporal control to the myogenic process. In this context, previous work showed that Pax7 is ubiquitinated by the E3 ligase neural precursor cell-expressed developmentally downregulated protein 4 and interacts with several proteins related to the ubiquitin-proteasome system, including the deubiquitinase ubiquitin-specific protease 7 (USP7). Although USP7 functions in diverse cellular contexts, its role(s) during myogenesis remains poorly explored. Here, we show that USP7 is transiently expressed in adult muscle progenitors, correlating with the onset of myogenin expression, while it is downregulated in newly formed myotubes/myofibers. Acute inhibition of USP7 activity upon muscle injury results in persistent expression of early regeneration markers and a significant reduction in the diameter of regenerating myofibers. At the molecular level, USP7 downregulation or pharmacological inhibition impairs muscle differentiation by affecting myogenin stability. Co-immunoprecipitation and in vitro activity assays indicate that myogenin is a novel USP7 target for deubiquitination. These results suggest that USP7 regulates SC myogenic progression by enhancing myogenin stability.
Sigma-Aldrich
MISSION® esiRNA, targeting human USP7
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)