Merck
  • Mesenchymal stem cells ameliorate lipid metabolism through reducing mitochondrial damage of hepatocytes in the treatment of post-hepatectomy liver failure.
Mesenchymal stem cells ameliorate lipid metabolism through reducing mitochondrial damage of hepatocytes in the treatment of post-hepatectomy liver failure.
Cell death & disease (2021-01-23)
Jing-Lin Wang, Hao-Ran Ding, Chen-Yan Pan, Xiao-Lei Shi, Hao-Zhen Ren
ABSTRACT
Hepatectomy is an effective therapeutic strategy for many benign and malignant liver diseases, while the complexity of liver anatomy and the difficulty of operation lead to complications after hepatectomy. Among them, post-hepatectomy liver failure (PHLF) is the main factor threatening the life of patients. At present, liver transplantation is an effective approach for PHLF. However, the application of liver transplantation has been largely limited due to the shortage of donors and the high cost of such operation. Therefore, it is urgently necessary to develop a new treatment for PHLF. Mesenchymal stem cells (MSCs) have become a new treatment regimen for liver diseases because of their easy access and low immunogenicity. Our study found that there were some subtle connections between MSCs and liver lipid metabolism in the PHLF model. We used MSC transplantation to treat PHLF induced by 90% hepatectomy. MSC transplantation could restore the mitochondrial function, promote the β-oxidation of fatty acid (FA), and reduce the lipid accumulation of hepatocytes. In addition, interleukin 10 (IL-10), a cytokine with immunoregulatory function, had an important role in lipid metabolism. We also found that MSCs transplantation activated the mammalian target of rapamycin (mTOR) pathway. Therefore, we explored the relationship between mitochondrial damage and lipid metabolism abnormality or PHLF. MSCs improved mitochondrial function and corrected abnormal lipid metabolism by affecting the mTOR pathway in the treatment of PHLF. Collectively, MSC transplantation could be used as a potential treatment for PHLF.
Sigma-Aldrich
Anti-CPT1A Antibody, serum, from rabbit
Sigma-Aldrich
Anti-Uncoupling Protein 1 Antibody, Chemicon®, from rabbit