Merck
  • Antioxidant and immunomodulatory activity induced by stevioside in liver damage: In vivo, in vitro and in silico assays.
Antioxidant and immunomodulatory activity induced by stevioside in liver damage: In vivo, in vitro and in silico assays.
Life sciences (2019-03-21)
Sael Casas-Grajales, Erika Ramos-Tovar, Esmeralda Chávez-Estrada, Diana Alvarez-Suarez, Erika Hernández-Aquino, Karina Reyes-Gordillo, Carlos M Cerda-García-Rojas, Javier Camacho, Víctor Tsutsumi, M Raj Lakshman, Pablo Muriel
ABSTRACT
Stevioside is a diterpenoid obtained from the leaves of Stevia rebaudiana (Bertoni) that exhibits antioxidant, antifibrotic, antiglycemic and anticancer properties. Therefore, we aimed to study whether stevioside has beneficial effects in liver injury induced by long-term thioacetamide (TAA) administration and investigated the possible underlying molecular mechanism using in vivo, in vitro and in silico approaches. Liver injury was induced in male Wistar rats by TAA administration (200 mg/kg), intraperitoneally, three times per week. Rats received saline or stevioside (20 mg/kg) twice daily intraperitoneally. In addition, cocultures were incubated with either lipopolysaccharide or ethanol. Liver injury, antioxidant and immunological responses were evaluated. Chronic TAA administration induced significant liver damage. In addition, TAA upregulated the protein expression of nuclear factor (NF)-κB, thus increasing the expression of proinflammatory cytokines and decreasing the antioxidant capacity of the liver through downregulation of nuclear erythroid factor 2 (Nrf2). Notably, stevioside administration prevented all of these changes. In vitro, stevioside prevented the upregulation of several genes implicated in liver inflammation when cocultured cells were incubated with lipopolysaccharide or ethanol. In silico assays using tumor necrosis factor receptor (TNFR)-1 and Toll-like receptor (TLR)-4-MD2 demonstrated that stevioside docks with TNFR1 and TLR4-MD2, thus promoting an antagonistic action against this proinflammatory mediator. Collectively, these data suggest that stevioside prevented liver damage through antioxidant activity by upregulating Nrf2 and immunomodulatory activity by blocking NF-κB signaling.
Sigma-Aldrich
Anti-NFκB Antibody, p65 subunit, active subunit, clone 12H11, clone 12H11, Chemicon®, from mouse
Sigma-Aldrich
Anti-MMP-13 Antibody, clone LIPCO IID1, clone LIPCO IID1, Chemicon®, from mouse