- Targeted disruption of Mib2 causes exencephaly with a variable penetrance.
Targeted disruption of Mib2 causes exencephaly with a variable penetrance.
Genesis (New York, N.Y. : 2000) (2007-11-08)
Jiang I Wu, Rashmi Rajendra, Julius C Barsi, Larissa Durfee, Eva Benito, Guang Gao, Mariam Kuruvilla, Radmila Hrdlicková, Andrew S Liss, Karen Artzt
PMID17987667
ABSTRACT
Mib1 and Mib2 ubiquitin ligases are very similar in their domain construction. They partake in the Notch signaling pathway by ubiquitinating the Notch receptors Delta and Jagged prior to endocytosis. We have created a targeted mutation of Mib2 and show that its phenotype is a variable penetrance, failure to close the cranial neural tube. The penetrance depends on the genetic background but it appears that Mib2 is not completely essential in mouse development.