Merck
  • TM4SF19 aggravates LPS-induced attenuation of vascular endothelial cell adherens junctions by suppressing VE-cadherin expression.
TM4SF19 aggravates LPS-induced attenuation of vascular endothelial cell adherens junctions by suppressing VE-cadherin expression.
Biochemical and biophysical research communications (2020-10-17)
Li Ding, Li-Min Li, Bing Hu, Jia-Li Wang, Yuan-Bin Lu, Ru-Yi Zhang, Xin He, Chao Shi, Li-Mei Wu, Chang-Meng Wu, Biao Yang, Lei Zheng, Bao-Hong Ping, Yan-Wei Hu, Qian Wang
ABSTRACT
Atherosclerosis is a chronic vascular inflammatory disease that initially starts from an arterial intima lesion and endothelial barrier dysfunction. The purpose of this study was to investigate the role of TM4SF19, a recently identified member of the transmembrane 4L six superfamily, in vascular endothelial cell adherens junctions. We found TM4SF19 expression was significantly increased in atherosclerotic plaques and sera of patients with coronary heart disease (CHD) compared with healthy people by immunohistochemistry and ELISA. In vitro, human umbilical vein endothelial cells (HUVECs) were stimulated by lipopolysaccharides (LPS). TM4SF19 and VE-cadherin expression as well as cell adherens junctions were assessed. Additionally, LPS could upregulate TM4SF19 expression and downregulate VE-cadherin expression in HUVECs in a concentration dependent manner. Overexpression of TM4SF19 substantially aggravated LPS-induced reduction of VE-cadherin expression and attenuation of vascular endothelial cell adherens junctions. However, both the decreased VE-cadherin expression and weakened cell adherens junctions induced by LPS could be dramatically reversed when the expression of TM4SF19 was depressed. This study is the first to reveal the effect of TM4SF19 on endothelial cell adherens junctions. Meanwhile, our results also provide novel therapeutic strategies for atherosclerotic diseases.
Sigma-Aldrich
Triton X-100, laboratory grade
Pricing and availability is not currently available.