Merck
  • Tailored gatifloxacin Pluronic® F-68-loaded contact lens: Addressing the issue of transmittance and swelling.
Tailored gatifloxacin Pluronic® F-68-loaded contact lens: Addressing the issue of transmittance and swelling.
International journal of pharmaceutics (2020-04-03)
Furqan A Maulvi, Riya J Parmar, Ankita R Desai, Ditixa M Desai, Manish R Shukla, Ketan M Ranch, Shailesh A Shah, Dinesh O Shah
ABSTRACT
Loading of gatifloxacin in contact lenses affects critical lens properties (optical and swelling) owing to drug precipitation in the contact lens matrix. The presence of Pluronic® F-68 in the packaging solution creates in-situ micelles in the contact lens to dissolve gatifloxacin precipitates and provide sustained drug release. The micelles further improved the drug uptake from the drug-packaging solution to create an equilibrium of drug between the lens matrix and the packaging solution. In this study, we optimized gatifloxacin-pluronic-loaded contact lenses to achieve the desired optical transmittance, swelling, and gatifloxacin loading capacity as well as sustained drug delivery. Optimization of gatifloxacin-pluronic-loaded contact lens was carried out using a 32 factorial design by tailoring the concentration of Pluronic® F-68 in the packaging solution (X1) and the amount of gatifloxacin in the monomer solution (X2) to achieve the desired lens properties. The optimized batch (X1 = 0.3%w/v and X2 = 0.3%w/v) showed an optical transmittance of 92.84%, swelling of 92.36% and gatifloxacin loading capacity of 92.56 μg. The in vitro flux data of the optimized batch (GT-Pl-CL) showed sustained release up to 72 h, whereas soaked contact lenses (SM-CL) and direct gatifloxacin-loaded contact lenses (DL-CL) showed a sustained release up to 48 h. The in vivo gatifloxacin release data for rabbit tear fluid showed sustained release with a high gatifloxacin level for the GT-Pl-CL lens in comparison to the SM-CL and the eye drop solution. This study demonstrates the application of the 32 full factorial design to optimize gatifloxacin-pluronic-loaded contact lenses to achieve the desired optical transmittance, swelling, and drug loading capacity.
Sigma-Aldrich
1-Hydroxycyclohexyl phenyl ketone, 99%