Merck
  • The VEGFR Inhibitor Cediranib Improves the Efficacy of Fractionated Radiotherapy in a Colorectal Cancer Xenograft Model.
The VEGFR Inhibitor Cediranib Improves the Efficacy of Fractionated Radiotherapy in a Colorectal Cancer Xenograft Model.
European surgical research. Europaische chirurgische Forschung. Recherches chirurgicales europeennes (2016-12-22)
Elodie Melsens, Bert Verberckmoes, Natacha Rosseel, Christian Vanhove, Benedicte Descamps, Piet Pattyn, Wim Ceelen
ABSTRACT
Radiotherapy (RT) increases local tumor control in locally advanced rectal cancer, but complete histological response is seen in only a minority of cases. Antiangiogenic therapy has been proposed to improve RT efficacy by "normalizing" the tumor microvasculature. Here, we examined whether cediranib, a pan-vascular endothelial growth factor (VEGF) receptor tyrosine kinase inhibitor, improves microvascular function and tumor control in combination with RT in a mouse colorectal cancer (CRC) model. CRC xenografts (HT29) were grown subcutaneously in mice. Animals were treated for 5 consecutive days with vehicle, RT (1.8 Gy daily), cediranib (6 mg/kg po), or combined therapy (cediranib 2 h prior to radiation). Tumor volume was measured with calipers. Vascular changes were analyzed by dynamic contrast-enhanced MRI, oxygenation and interstitial fluid pressure probes and histology. To investigate vascular changes more in detail, a second set of mice were fitted with titanium dorsal skinfold window chambers, wherein a HT29 tumor cell suspension was injected. In vivo fluorescence microscopy was performed before and after treatment (same treatment protocol). In vivo microscopy analyses showed that VEGFR inhibition with cediranib led to a "normalization" of the vessel wall, with decreased microvessel permeability (p < 0.0001) and tortuosity (p < 0.01), and a trend to decreased vessel diameters. This seemed to lead to lower tumor hypoxia rates in the cediranib and combination groups compared to the control and RT groups. This led to an increased tumor control in the combination group compared to controls or monotherapy (p < 0.0001). The combination of RT with cediranib enhances tumor control in a CRC xenograft mouse model. Microvascular analyses suggest that cediranib leads to vascular normalization and improved oxygenation.
Product Description
Roche
In Situ Cell Death Detection Kit, Fluorescein, sufficient for ≤50 tests, suitable for detection
Sigma-Aldrich
Cediranib, ≥98% (HPLC)