Merck
  • C-reactive protein specifically enhances platelet-activating factor-induced inflammatory activity in vivo.
C-reactive protein specifically enhances platelet-activating factor-induced inflammatory activity in vivo.
European journal of pharmacology (2014-12-03)
Akira Sato, Keitaro Oe, Hikaru Yamanaka, Izumi Yokoyama, Keiichi Ebina
ABSTRACT
Platelet-activating factor (PAF) is a potent lipid mediator that is implicated in numerous inflammatory diseases. C-reactive protein (CRP) is an acute-phase plasma protein that increases rapidly and dramatically in response to inflammation. In this study, we investigated the effect of the interaction between CRP and PAF on inflammatory responses in vivo. From binding analysis using a time-resolved fluorometric assay, CRP bound to PAF and its precursor/metabolite lyso-PAF in a concentration-dependent manner. In addition, CRP bound to several phospholipids containing lysophosphatidylcholine, which bears structural resemblance to PAF and lyso-PAF, sphingosylphosphorylcholine, and lysophosphatidylethanolamine more readily than to lysophosphatidic acid and lysophosphatidylserine. In in vivo experiments using a rat model of hind paw oedema, CRP increased PAF-induced rat paw oedema in a dose-dependent manner, without causing the oedema itself, but it did not increase histamine and serotonin-induced paw oedema. Furthermore, the receptor for CRP, lectin-like oxidized low-density lipoprotein receptor 1 was not involved in the increase in PAF-induced inflammatory responses caused by CRP. These results indicate that CRP can specifically enhance PAF-induced inflammatory activity through binding to PAF and lyso-PAF. Therefore, CRP may accelerate the pathogenesis of numerous inflammatory diseases caused by PAF.
Product Description
Avanti
C16-18:1 PC, Avanti Research - A Croda Brand 878112C
Avanti
C16-02:0 PC, Avanti Research - A Croda Brand 878110P, powder