Merck
  • Isomerization of aldehyde-2,4-dinitrophenylhydrazone derivatives and validation of high-performance liquid chromatographic analysis.
Isomerization of aldehyde-2,4-dinitrophenylhydrazone derivatives and validation of high-performance liquid chromatographic analysis.
Journal of chromatography. A (2003-07-02)
Shigehisa Uchiyama, Masanori Ando, Shohei Aoyagi
ABSTRACT
The most widely used method for qualitative and quantitative analysis of carbonyl compounds is the 2,4-dinitrophenylhydrazine method through the formation of 2,4-dinitrophenylhydrazone derivatives. However, this method may cause an analytical error because 2,4-dinitrophenylhydrazones have both E- and Z-stereoisomers. Purified aldehyde-2,4-dinitrophenylhydrazone demonstrated only the E-isomer. However under UV irradiation and the addition of acid, both E- and Z-isomers were seen. The spectral patterns of Z-isomers were different from those of E-isomers and the absorption maximum wavelengths were shifted towards shorter wavelengths by 5-8 nm. An equilibrium Z/E isomer ratio was observed in 0.02-0.2% (v/v) phosphoric acid solutions. In the case of acetaldehyde- and propanal-2,4-dinitrophenylhydrazones, the equilibrium Z/E isomer ratios were 0.32 and 0.14, respectively. However, when irradiated with ultraviolet light at 364 nm, the isomer ratios were increased beyond this constant ratio and reached 0.55 and 0.33, respectively. Zero-order rates for decreases of aldehyde derivatives were observed under UV irradiation (364 nm), however the decreases of concentration were not observed in phosphoric acid solutions. The best method for the determination of aldehyde-2,4-dinitrophenylhydrazones by HPLC is to add phosphoric acid to both the sample and the standard solution, to form a 0.02-1% acid solution.