Merck
  • Benzalkonium ion sorption to peat and clays: Relative contributions of ion exchange and van der Waals interactions.
Benzalkonium ion sorption to peat and clays: Relative contributions of ion exchange and van der Waals interactions.
Chemosphere (2020-01-25)
Mlamuli Ndabambi, Jung-Hwan Kwon
ABSTRACT
Due to their use in various domestic and industrial formulations, benzalkonium compounds have been isolated in many environmental matrices. Sorption to soil components has been shown to play a key role in their environmental fate. Whereas sorption of benzalkonium compounds to soils is attributed to cation exchange and van der Waals forces, the relative contributions of these two mechanisms at environmental levels have not been clearly defined. In this study a previously reported algal toxicity assay-based method was employed to determine the distribution coefficients (Kd) of benzalkonium compounds between water and soil components, at environmental concentrations. Cation exchange capacity corrected Kd values for organic matter and clays were all within one order of magnitude. This implies that ion exchange is the dominant mechanism of sorption for benzalkonium compounds. When the sorption data were used to compute sorption energies for four homologues of benzalkonium ions, the magnitude of the free energy change of sorption increased with size of the molecule. The increase in sorption energy could be partly explained by increased energy of hydration with addition of methylene groups to the alkyl chain. A model that predicts sorption coefficients of benzalkonium compounds to soils using organic carbon content and cation exchange capacity was also defined. When tested using an artificial soil, the model estimates were all within one order of magnitude of the experimental values.
Sigma-Aldrich
Benzyldimethyldodecylammonium chloride, ≥99.0% (AT)
Sigma-Aldrich
Benzyldimethyloctylammonium chloride, ≥96.0% (AT)